TIG Welding Gauntlets
Product Details & Documents
The TIG Ultra Welding Gauntlet offers fantastic dexterity with a goat leather palm and excellent burn resistance due to the split leather backing. Designed for the professional TIG welder with aramid stitching for ultra durability.
- EN12477 Type A welding protection
- Premium quality leather welding gauntlet
- The ultimate in dexterity for welders
- Made from high quality durable goat leather
Ideal for welding and metal handling, these gloves feature reinforced Aramid stitching for maximum durability and burn protection.
Colour: Brown
Certifications: EN ISO 21420: 2020 Dexterity 2, EN 388: 2016 + A1: 2018 3243X, EN 12477: 2001 Type A, EN 407: 2020 422X31. ANSI/ISEA 105: 2016 CUT Level A2. CE certified.
Documents
TDS A521 - TIG Welding Gauntlet
Datasheet
Specifications
- Glove size:XL
- Colour:Brown
- Seq:02
- Cat. No.:PRTWA521BRRXL
- Latex / latex-free:Latex-free
- Disposable / reusable:Reusable
- Material:Leather/polyester
- Powdered / powder-free:Powder-free
- Sterility:Non sterile
- Ambidextrous / hand-specific:Hand-specific
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



