Order Entry
Poland
ContactUsLinkComponent
FITC-Labeled Human Mesothelin (296-580), His Tag (MSN-HF2H3) is produced via conjugation of FITC to Human Mesothelin (296-580), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Mesothelin (296-580), 1 * 25 µG
  ACRBMSN-HF2H3-25UG
FITC-Labeled Human Mesothelin (296-580), His Tag (MSN-HF2H3) is produced via conjugation of FITC to Human Mesothelin (296-580), His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Mesothelin (296-580), 1 * 25 µG
  ACRBMSN-HF2H3-25UG
Frequently Bought Together