Order Entry
Poland
ContactUsLinkComponent
Biotinylated Mouse BAFF, Tag Free, primary amine labeling (BAF-M8219) is expressed from human 293 cells (HEK293). It contains AA Ala 127 - Leu 309 (Accession # Q9WU72-1). 1 * 200 µG
Catalog # ACRBBAF-M8219200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse BAFF, Tag Free, primary amine labeling (BAF-M8219) is expressed from human 293 cells (HEK293). It contains AA Ala 127 - Leu 309 (Accession # Q9WU72-1). 1 * 200 µG
Catalog # ACRBBAF-M8219200UG
Supplier:  ACRO Biosystems
Frequently Bought Together