Order Entry
Poland
ContactUsLinkComponent
PE-Labeled Human VEGF R2, His Tag (KDR-HP227) is produced via site-specific conjugation of PE to Human VEGF R2, His Tag under optima l conditions with a proprietary technology. Human VEGF R2, His Tag is expressed from human 293 cells (HEK293). It con 1 * 200 Tests
Catalog # ACRBKDR-HP227-200
Supplier:  ACRO Biosystems
PE-Labeled Human VEGF R2, His Tag (KDR-HP227) is produced via site-specific conjugation of PE to Human VEGF R2, His Tag under optima l conditions with a proprietary technology. Human VEGF R2, His Tag is expressed from human 293 cells (HEK293). It con 1 * 200 Tests
Catalog # ACRBKDR-HP227-200
Supplier:  ACRO Biosystems
Frequently Bought Together