0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Mouse N-APP, His Tag (APP-M52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 18 - Val 286 (Accession # P12023-1). 1 * 100 µG
ACRBAPP-M52H3100UG
:
ACRO Biosystems
Mouse N-APP, His Tag (APP-M52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 18 - Val 286 (Accession # P12023-1). 1 * 100 µG
ACRBAPP-M52H3100UG
:
ACRO Biosystems
Frequently Bought Together
Serological pipettes, Stripette™
: Corning
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
: BIOCISION
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
Sterilin™ Quickstart Universal Containers, PS, 30 ml
: STERILIN
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
: VWR Collection
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Optical bottom plates, 96-well, Nunclon™Δ
: Thermo Fisher Scientific
VWR®, Cryo coder cap inserts
: VWR Collection
Cell culture storage bottles, polystyrene, Corning®
: Corning
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection