Order Entry
Poland
ContactUsLinkComponent
Human NKp46, His Tag (NC1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Arg 258 (Accession # O76036-1). 1 * 100 µG
Catalog # ACRBNC1-H52H6100UG
Supplier:  ACRO Biosystems
Human NKp46, His Tag (NC1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Arg 258 (Accession # O76036-1). 1 * 100 µG
Catalog # ACRBNC1-H52H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together