Order Entry
Poland
ContactUsLinkComponent
APC-Labeled Human Mesothelin (296-580), His Tag (MSN-HA2H7) is produced via conjugation of APC to Human Mesothelin (296-580), His Ta g with a new generation site-specific technology under Star Staining labeling platform. Human Mesothelin (296-580), H 1 * 25 Tests
Catalog # ACRBMSN-HA2H7-25
Supplier:  ACRO Biosystems
APC-Labeled Human Mesothelin (296-580), His Tag (MSN-HA2H7) is produced via conjugation of APC to Human Mesothelin (296-580), His Ta g with a new generation site-specific technology under Star Staining labeling platform. Human Mesothelin (296-580), H 1 * 25 Tests
Catalog # ACRBMSN-HA2H7-25
Supplier:  ACRO Biosystems
Frequently Bought Together