Order Entry
Poland
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (BA.2.12.1/Omicron) (SPN-C82Es) is expressed from human 293 cells (HEK293). It cont ains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, G142D, V213G, G339D, S371F, S373P, S375F, 1 * 25 µG
Catalog # ACRBSPN-C82ES-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (BA.2.12.1/Omicron) (SPN-C82Es) is expressed from human 293 cells (HEK293). It cont ains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, G142D, V213G, G339D, S371F, S373P, S375F, 1 * 25 µG
Catalog # ACRBSPN-C82ES-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together