Order Entry
Poland
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike NTD, His,Avitag (BA.2/Omicron) (SPD-C82H5) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, G142D, V213G)). The spike mutations are identified 1 * 25 µG
Catalog # ACRBSPD-C82H5-25UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike NTD, His,Avitag (BA.2/Omicron) (SPD-C82H5) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, G142D, V213G)). The spike mutations are identified 1 * 25 µG
Catalog # ACRBSPD-C82H5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together