Biotinylated SARS-CoV-2 Spike S2 protein, His,Avitag™ (BA.2/Omicron) (S2N-C82E4) is expressed from human 293 cells (HEK293). It cont ains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (N764K, D796Y, Q954H, N969K, F817P, A892P, A899P, A942P, K986P, V9 1 * 200 µG
Catalog # ACRBS2N-C82E4200UG
Supplier: ACRO Biosystems
Frequently Bought Together