Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 S2 protein, His Tag (BA.1/Omicron) (S2N-C52Hf) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1 213 (Accession # QHD43416.1 (N764K, D796Y, N856K, Q954H, N969K, L981F, F817P, A892P, A899P, A942P, K986P, V987P)). Th 1 * 1 mg
Catalog # ACRBS2N-C52HF-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S2 protein, His Tag (BA.1/Omicron) (S2N-C52Hf) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1 213 (Accession # QHD43416.1 (N764K, D796Y, N856K, Q954H, N969K, L981F, F817P, A892P, A899P, A942P, K986P, V987P)). Th 1 * 1 mg
Catalog # ACRBS2N-C52HF-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together