Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (BA.2/Omicron) (SPN-C522b) is expressed from human 293 cells (HEK293). The spike mutations are iden tified on the SARS-CoV-2 Omicron variant (Pango lineage: BA.2 GISAID clade: GRA Nextstrain clade: 21L). 1 * 10 mg
Catalog # ACRBSPN-C522B-10MG
Dostawca:  ACRO Biosystems
SARS-CoV-2 Spike Trimer, His Tag (BA.2/Omicron) (SPN-C522b) is expressed from human 293 cells (HEK293). The spike mutations are iden tified on the SARS-CoV-2 Omicron variant (Pango lineage: BA.2 GISAID clade: GRA Nextstrain clade: 21L). 1 * 10 mg
Catalog # ACRBSPN-C522B-10MG
Dostawca:  ACRO Biosystems
Frequently Bought Together