Order Entry
Poland
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag™ (BA.2/Omicron) (SPN-C82Er) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, G142D, V213G, G339D, S371F, S373P, S375F, T37 1 * 200 µG
Catalog # ACRBSPN-C82ER200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag™ (BA.2/Omicron) (SPN-C82Er) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, G142D, V213G, G339D, S371F, S373P, S375F, T37 1 * 200 µG
Catalog # ACRBSPN-C82ER200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together