Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (BA.1+L452R/Omicron) (SPD-C522k) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339D, S371L, S373P, S375F, K417N, N440K, G446S, L452R, S477N, T478K, E484A, Q493R, G 1 * 100 µG
  ACRBSPD-C522K100UG
SARS-CoV-2 Spike RBD, His Tag (BA.1+L452R/Omicron) (SPD-C522k) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339D, S371L, S373P, S375F, K417N, N440K, G446S, L452R, S477N, T478K, E484A, Q493R, G 1 * 100 µG
  ACRBSPD-C522K100UG
Frequently Bought Together