Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (XD/BA.1 x AY.4 recombinant) (SPN-C5226) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19R, A27S, T95I, G142D, EFR156G, NL211I, INS214EPE, G339D, S371L, S373P, 1 * 500 µG
  ACRBSPN-C5226500UG
SARS-CoV-2 Spike Trimer, His Tag (XD/BA.1 x AY.4 recombinant) (SPN-C5226) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19R, A27S, T95I, G142D, EFR156G, NL211I, INS214EPE, G339D, S371L, S373P, 1 * 500 µG
  ACRBSPN-C5226500UG
Frequently Bought Together