Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein, His Tag (BA.3*(S33V)/Omicron) (NUN-C52Hv) is expressed from human 293 cells (HEK293). It contains A A Met 1 - Ala 419 (Accession # QHO62115.1 (P13L, GER 30-32del, S33V, R203K, G204R, S413R)). The mutations (P13L, GER 1 * 100 µG
Catalog # ACRBNUN-C52HV100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein, His Tag (BA.3*(S33V)/Omicron) (NUN-C52Hv) is expressed from human 293 cells (HEK293). It contains A A Met 1 - Ala 419 (Accession # QHO62115.1 (P13L, GER 30-32del, S33V, R203K, G204R, S413R)). The mutations (P13L, GER 1 * 100 µG
Catalog # ACRBNUN-C52HV100UG
Supplier:  ACRO Biosystems
Frequently Bought Together