Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (BA.3/Omicron) (SPD-C522i) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 53 7 (Accession # QHD43416.1 (G339D,S371F,S373P,S375F,D405N,K417N,N440K,G446S,S477N,T478K,E484A,Q493R,Q498R,N501Y,Y505H) 1 * 1 mg
Catalog # ACRBSPD-C522I-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike RBD, His Tag (BA.3/Omicron) (SPD-C522i) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 53 7 (Accession # QHD43416.1 (G339D,S371F,S373P,S375F,D405N,K417N,N440K,G446S,S477N,T478K,E484A,Q493R,Q498R,N501Y,Y505H) 1 * 1 mg
Catalog # ACRBSPD-C522I-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together