Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (BA.3/Omicron) (SPN-C5225) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (A67V,H69del,V70del,T95I,G142D,V143del,Y144del,Y145del,N211del,L212I,G339D,S371F,S373P,S3 1 * 500 µG
Catalog # ACRBSPN-C5225500UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike Trimer, His Tag (BA.3/Omicron) (SPN-C5225) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (A67V,H69del,V70del,T95I,G142D,V143del,Y144del,Y145del,N211del,L212I,G339D,S371F,S373P,S3 1 * 500 µG
Catalog # ACRBSPN-C5225500UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together