Order Entry
Poland
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (BA.2/Omicron) (SPD-C82Eq) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, S477N, T478 1 * 25 µG
Catalog # ACRBSPD-C82EQ-25UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (BA.2/Omicron) (SPD-C82Eq) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, S477N, T478 1 * 25 µG
Catalog # ACRBSPD-C82EQ-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together