Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Spike S1, His Tag (BA.2/Omicron) (S1N-C52Hv) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1(T19I, LPP24-26del, A27S, G142D, V213G, G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, 1 * 1 mg
Catalog # ACRBS1N-C52HV-1MG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike S1, His Tag (BA.2/Omicron) (S1N-C52Hv) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1(T19I, LPP24-26del, A27S, G142D, V213G, G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, 1 * 1 mg
Catalog # ACRBS1N-C52HV-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together