Order Entry
Poland
ContactUsLinkComponent
Epstein-Barr virus Envelope Glycoprotein B (gB), His Tag (GLB-E52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 685 (Accession # P03188 (W112H, Y113R, R428G, R429S, R430G, R431S, R432G, W193R, L194V, I195E, W196A)). 1 * 100 µG
Catalog # ACRBGLB-E52H3100UG
Supplier:  ACROBIOSYSTEMS
Epstein-Barr virus Envelope Glycoprotein B (gB), His Tag (GLB-E52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 685 (Accession # P03188 (W112H, Y113R, R428G, R429S, R430G, R431S, R432G, W193R, L194V, I195E, W196A)). 1 * 100 µG
Catalog # ACRBGLB-E52H3100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together