Order Entry
Poland
ContactUsLinkComponent
HCMV Glycoprotein B (gB) Protein (Strain AD169), His Tag is the ectodomain of HCMV (strain AD169) glycoprotein B (gB) which contains AA Ser 25 – Phe 706 (Accession # P06473 (Y155G, I156H, Y157R, Y206H, W240A, L241T, Y242H, R456S, R459S)). The recomb 1 * 50 µG
Catalog # ACRBCMB-V52H4-50UG
Dostawca:  ACRO Biosystems
HCMV Glycoprotein B (gB) Protein (Strain AD169), His Tag is the ectodomain of HCMV (strain AD169) glycoprotein B (gB) which contains AA Ser 25 – Phe 706 (Accession # P06473 (Y155G, I156H, Y157R, Y206H, W240A, L241T, Y242H, R456S, R459S)). The recomb 1 * 50 µG
Catalog # ACRBCMB-V52H4-50UG
Dostawca:  ACRO Biosystems
Frequently Bought Together