Order Entry
Poland
ContactUsLinkComponent
Nipah virus Glycoprotein, His Tag (GLN-N52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 71 - Thr 602 (Accession # Q9IH62-1). 1 * 50 µG
  ACRBGLN-N52H3-50UG
Nipah virus Glycoprotein, His Tag (GLN-N52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 71 - Thr 602 (Accession # Q9IH62-1). 1 * 50 µG
  ACRBGLN-N52H3-50UG
Frequently Bought Together