Order Entry
Poland
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (SPD-C82Em) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 5 37 (Accession # QHD43416.1 (R346K, E484K, N501Y)). The mutations R346K, E484K, N501Y were identified in the SARS-CoV- 1 * 25 µG
Catalog # ACRBSPD-C82EM-25UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (SPD-C82Em) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 5 37 (Accession # QHD43416.1 (R346K, E484K, N501Y)). The mutations R346K, E484K, N501Y were identified in the SARS-CoV- 1 * 25 µG
Catalog # ACRBSPD-C82EM-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together