Order Entry
Poland
ContactUsLinkComponent
Hendra virus Glycoprotein, His Tag (GLN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 71 - Ser 604 (Accessio n # O89343-1). 1 * 50 µG
Catalog # ACRBGLN-H52H3-50UG
Supplier:  ACRO Biosystems
Hendra virus Glycoprotein, His Tag (GLN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 71 - Ser 604 (Accessio n # O89343-1). 1 * 50 µG
Catalog # ACRBGLN-H52H3-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together