Order Entry
Poland
ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (SPD-C5221) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (R346K, E484K, N501Y)). The mutations R346K, E484K, N501Y were identified in the SARS-CoV-2 Mu variant (Pan 1 * 100 µG
Catalog # ACRBSPD-C5221100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike RBD, His Tag (SPD-C5221) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (R346K, E484K, N501Y)). The mutations R346K, E484K, N501Y were identified in the SARS-CoV-2 Mu variant (Pan 1 * 100 µG
Catalog # ACRBSPD-C5221100UG
Supplier:  ACRO Biosystems
Frequently Bought Together