Order Entry
Poland
ContactUsLinkComponent
Rhesus macaque CD117, Fc Tag (CD7-R5259) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 520 (Accession # NP _001253024.1). 1 * 100 µG
Catalog # ACRBCD7-R5259100UG
Supplier:  ACRO Biosystems
Rhesus macaque CD117, Fc Tag (CD7-R5259) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 520 (Accession # NP _001253024.1). 1 * 100 µG
Catalog # ACRBCD7-R5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together