Order Entry
Poland
ContactUsLinkComponent
Human IL-2 Protein, Fc Tag (IL2-H5269) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession # P605 68-1). 1 * 1 mg
Catalog # ACRBIL2-H5269-1MG
Supplier:  ACRO Biosystems
Human IL-2 Protein, Fc Tag (IL2-H5269) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession # P605 68-1). 1 * 1 mg
Catalog # ACRBIL2-H5269-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together