Order Entry
Poland
ContactUsLinkComponent
Human Fc gamma RIIIA / CD16a (F176, S197P), His Tag (CDA-H52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (F176, S197P) (Accession # P08637-1 (F176, S197P)). 1 * 100 µG
Catalog # ACRBCDA-H52H6100UG
Supplier:  ACRO Biosystems
Human Fc gamma RIIIA / CD16a (F176, S197P), His Tag (CDA-H52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (F176, S197P) (Accession # P08637-1 (F176, S197P)). 1 * 100 µG
Catalog # ACRBCDA-H52H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together