Order Entry
Poland
ContactUsLinkComponent
Biotinylated Mouse IL-27 Protein, His,Avitag (IL7-M82E4) is expressed from human 293 cells (HEK293). It contains AA Tyr 19 - Pro 228 (IL-27B) & Phe 29 - Ser 234 (IL-27A) (Accession # O35228-1 (IL-27B) & Q8K3I6-1 (IL-27A)). 1 * 200 µG
Catalog # ACRBIL7-M82E4200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse IL-27 Protein, His,Avitag (IL7-M82E4) is expressed from human 293 cells (HEK293). It contains AA Tyr 19 - Pro 228 (IL-27B) & Phe 29 - Ser 234 (IL-27A) (Accession # O35228-1 (IL-27B) & Q8K3I6-1 (IL-27A)). 1 * 200 µG
Catalog # ACRBIL7-M82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together