Order Entry
Poland
ContactUsLinkComponent
Human ITGAD&ITGB2 Heterodimer Protein, His Tag&Tag Free (IT2-H53W6) is expressed from CHO cells. It contains AA Phe 18 - Asn 1099 (I TGAD) & Gln 23 - Asn 700 (ITGB2) (Accession # Q13349-1 (ITGAD) & P05107-1 (ITGB2)). 1 * 1 mg
  ACRBIT2-H53W6-1MG
Human ITGAD&ITGB2 Heterodimer Protein, His Tag&Tag Free (IT2-H53W6) is expressed from CHO cells. It contains AA Phe 18 - Asn 1099 (I TGAD) & Gln 23 - Asn 700 (ITGB2) (Accession # Q13349-1 (ITGAD) & P05107-1 (ITGB2)). 1 * 1 mg
  ACRBIT2-H53W6-1MG
Frequently Bought Together