Order Entry
Poland
ContactUsLinkComponent
ActiveMax® Human TGFB1, Tag Free (TG1-H4212) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # P01137-1). 1 * 10 µG
Catalog # ACRBTG1-H4212-10UG
Supplier:  ACRO Biosystems
ActiveMax® Human TGFB1, Tag Free (TG1-H4212) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # P01137-1). 1 * 10 µG
Catalog # ACRBTG1-H4212-10UG
Supplier:  ACRO Biosystems
Frequently Bought Together