Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human IL-1 RAcP, His,Avitag (ILP-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Glu 359 (Ac cession # Q9NPH3-1). 1 * 25 µG
Catalog # ACRBILP-H82E5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-1 RAcP, His,Avitag (ILP-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Glu 359 (Ac cession # Q9NPH3-1). 1 * 25 µG
Catalog # ACRBILP-H82E5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together