Order Entry
Poland
ContactUsLinkComponent
Human CD131, Fc Tag (CD1-H5256) is expressed from human 293 cells (HEK293). It contains AA Trp 17 - Trp 443 (Accession # P32927-1). 1 * 1 mg
Catalog # ACRBCD1-H5256-1MG
Supplier:  ACRO Biosystems
Human CD131, Fc Tag (CD1-H5256) is expressed from human 293 cells (HEK293). It contains AA Trp 17 - Trp 443 (Accession # P32927-1). 1 * 1 mg
Catalog # ACRBCD1-H5256-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together