Order Entry
Poland
ContactUsLinkComponent
Human IGFBP-3, His Tag (IG3-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Lys 291 (Accession # P17936-1 ). 1 * 100 µG
Catalog # ACRBIG3-H52H9100UG
Supplier:  ACRO Biosystems
Human IGFBP-3, His Tag (IG3-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Lys 291 (Accession # P17936-1 ). 1 * 100 µG
Catalog # ACRBIG3-H52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together