Order Entry
Poland
ContactUsLinkComponent
Human LAMP5, His Tag (LA5-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Glu 235 (Accession # Q9UJQ1-1). 1 * 500 µG
Catalog # ACRBLA5-H52H9500UG
Supplier:  ACRO Biosystems
Human LAMP5, His Tag (LA5-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Glu 235 (Accession # Q9UJQ1-1). 1 * 500 µG
Catalog # ACRBLA5-H52H9500UG
Supplier:  ACRO Biosystems
Frequently Bought Together