Order Entry
Poland
ContactUsLinkComponent
Human IL-2RB&IL-2RA&IL-2RG, His Tag&Twin Strep Tag (ILG-H52W9) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A sp 239 (IL-2RB) & Glu 22 - Ser 212 (IL-2RA) & Leu 23 - Asn 254 (IL-2RG) (Accession # P14784-1 (IL-2RB) & P01589-1 (IL 1 * 1 mg
Catalog # ACRBILG-H52W9-1MG
Supplier:  ACRO Biosystems
Human IL-2RB&IL-2RA&IL-2RG, His Tag&Twin Strep Tag (ILG-H52W9) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A sp 239 (IL-2RB) & Glu 22 - Ser 212 (IL-2RA) & Leu 23 - Asn 254 (IL-2RG) (Accession # P14784-1 (IL-2RB) & P01589-1 (IL 1 * 1 mg
Catalog # ACRBILG-H52W9-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together