Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human IL-2RB&IL-2RA&IL-2RG, His,Avitag&Twin Strep Tag (ILG-H82W9) is expressed from human 293 cells (HEK293). It contai ns AA Ala 27 - Asp 239 (IL-2RB) & Glu 22 - Ser 212 (IL-2RA) & Leu 23 - Asn 254 (IL-2RG) (Accession # P14784-1 (IL-2RB 1 * 25 µG
  ACRBILG-H82W9-25UG
Biotinylated Human IL-2RB&IL-2RA&IL-2RG, His,Avitag&Twin Strep Tag (ILG-H82W9) is expressed from human 293 cells (HEK293). It contai ns AA Ala 27 - Asp 239 (IL-2RB) & Glu 22 - Ser 212 (IL-2RA) & Leu 23 - Asn 254 (IL-2RG) (Accession # P14784-1 (IL-2RB 1 * 25 µG
  ACRBILG-H82W9-25UG
Frequently Bought Together