Order Entry
Poland
ContactUsLinkComponent
Human FGF-3, His Tag (FG3-H51H9) is expressed from E.coli cells. It contains AA Ala 18 - His 239 (Accession # P11487-1). 1 * 100 µG
Catalog # ACRBFG3-H51H9100UG
Supplier:  ACRO Biosystems
Human FGF-3, His Tag (FG3-H51H9) is expressed from E.coli cells. It contains AA Ala 18 - His 239 (Accession # P11487-1). 1 * 100 µG
Catalog # ACRBFG3-H51H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together