Order Entry
Poland
ContactUsLinkComponent
Cynomolgus LRRC32&TGF-beta 1 Heterotrimer Protein, His Tag&Tag Free (GA1-C52W7) is expressed from human 293 cells (HEK293). It conta ins AA Ala 18 -Asn 627 (LRRC32) & Leu 30 - Ser 390 (TGF-beta 1) (Accession # A0A2K5X2X9-1 (LRRC32) & A0A2K5TJB2-1 (TG 1 * 100 µG
  ACRBGA1-C52W7100UG
Cynomolgus LRRC32&TGF-beta 1 Heterotrimer Protein, His Tag&Tag Free (GA1-C52W7) is expressed from human 293 cells (HEK293). It conta ins AA Ala 18 -Asn 627 (LRRC32) & Leu 30 - Ser 390 (TGF-beta 1) (Accession # A0A2K5X2X9-1 (LRRC32) & A0A2K5TJB2-1 (TG 1 * 100 µG
  ACRBGA1-C52W7100UG
Frequently Bought Together