Order Entry
Poland
ContactUsLinkComponent
Human IL-35, Fc Tag (IL5-H5259) is expressed from human 293 cells (HEK293). It contains AA Arg 21 - Lys 229 (IL-27 beta) & Arg 23 - Ser 219 (IL-12 alpha) (Accession # Q14213-1 (IL-27 beta) & P29459-1 (IL-12 alpha)). 1 * 100 µG
Catalog # ACRBIL5-H5259100UG
Supplier:  ACRO Biosystems
Human IL-35, Fc Tag (IL5-H5259) is expressed from human 293 cells (HEK293). It contains AA Arg 21 - Lys 229 (IL-27 beta) & Arg 23 - Ser 219 (IL-12 alpha) (Accession # Q14213-1 (IL-27 beta) & P29459-1 (IL-12 alpha)). 1 * 100 µG
Catalog # ACRBIL5-H5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together