Order Entry
Poland
ContactUsLinkComponent
Biotinylated Cynomolgus / Rhesus macaque 4-1BB, Avitag,His Tag (41B-C82E9) is expressed from human 293 cells (HEK293). It contains A A Leu 24 - Gln 186 (Accession # A9YYE7-1). 1 * 25 µG
Catalog # ACRB41B-C82E9-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus / Rhesus macaque 4-1BB, Avitag,His Tag (41B-C82E9) is expressed from human 293 cells (HEK293). It contains A A Leu 24 - Gln 186 (Accession # A9YYE7-1). 1 * 25 µG
Catalog # ACRB41B-C82E9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together