NITRIC ACID 0.1 MOL/L DILUT-IT 1 * 60 mL
Catalog # 4892.
Supplier: Avantor
Frequently Bought Together
1-Propanol, EMPLURA®, Supelco®
Supplier: Merck
Trifluoroacetic acid ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
N,N-Dimethylformamide, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
9-Fluorenylmethyl chloroformate (Fmoc chloride), Supelco®
Supplier: Merck
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
Supplier: PanReac AppliChem
Diethyl ether stabilised, pure analytical reagent
Supplier: CHEMPUR
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
4-(Methylamino)phenol sulphate 99%, extra pure
Supplier: Thermo Fisher Scientific
(±)-1,2-Propanediol, pure
Supplier: CHEMPUR