0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
EPREDIA SHANDON EA-50 STAIN 1 * 4 L
SHAN6766002
:
SHANDON
EPREDIA SHANDON EA-50 STAIN 1 * 4 L
SHAN6766002
:
SHANDON
Frequently Bought Together
Giemsa's stain solution for microscopy, Sigma-Aldrich®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test strips for peracetic acid, MQuant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
Supelco® Spectroquant® Test Kits, Sulphate
: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test kits, photometric, Nitrate, LCK cuvette tests
: Hach
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals