Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Microelectrode tin Microelectrode for ElectraSyn 2.0, 12 pcs. 1 * 12 items
Catalog # IKAA40004037
Supplier:
IKA
Expand image
Microelectrode tin Microelectrode for ElectraSyn 2.0, 12 pcs. 1 * 12 items
Catalog # IKAA40004037
Supplier:
IKA
Frequently Bought Together
VWR® pHenomenal® pH Electrodes
Supplier: VWR Collection
VWR® Spec-Wipe® 3, Cleanroom Wipes, Pre-Saturated
Supplier: VWR Collection
pH electrodes, SenTix®
Supplier: WTW
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
VWR®, Wipes, Pre-Saturated
Supplier: VWR Collection
pH electrodes, InLab® ISM
Supplier: Mettler - Toledo
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Cleanroom Wipes, 100% Polyester
Supplier: VWR Collection
VWR®, Bench Protectors
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Facial tissues, KLEENEX®
Supplier: KIMBERLY CLARK