STANDARD PRZEWODNO?CI.12,90MS/CM100ML 1 * 100 mL
Catalog # ELME12.90MS/100ML
Supplier: ELMETRON
Frequently Bought Together
VWR® ARISTAR®, Cobalt Standard Solution, for ICP, 1000 mg/L Co in dil. Nitric Acid (from Co)
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
UV/Visible standards, Supelco®
Supplier: Merck
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
Supplier: VWR Chemicals
NexION Setup Solution 1 ug/L 500mL 1 * 1 items
Supplier: PerkinElmer
VWR®, Colour standard solutions ASTM
Supplier: VWR Chemicals
Pharmaceutical secondary standard, CMIT/MIT, (CRM), Supelco®
Supplier: Merck
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond