ANTYMONYLU POTASU WINIAN 3HYDRAT 1 * 10 g
CHMP111485605.0010
: CHEMPUR
Frequently Bought Together
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Sodium dihydrogen phosphate ≥98%, GPR RECTAPUR®
: VWR Chemicals
Potassium iodide ≥99% ACS
: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Iron(III) nitrate nonahydrate ≥98% (metals basis)
: Thermo Fisher Scientific