2'-FLUORO O-FLUOROFENTANYL-13C6 HCL 1 * 0,5 mL
Catalog # F-092-0.5ML
Supplier: Merck
Frequently Bought Together
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
pH buffer solutions, colour coded
Supplier: Hach
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
Chloride Standard for IC 1 * 100 mL
Supplier: Merck
Conductivity standards, KCL
Supplier: Hach