6-PHENYL-2.3-DIHYDROIMIDAZO[2.1-B]THIAZO 1 * 25 mg
PROOMM0706.04-0025
Frequently Bought Together
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, ready to use
: WTW
Buffer solutions in sachets, perpHect® Pouches, Orion™
: Thermo Orion