VWR®, Density Standards, Certified Reference Materials, Referenced at 20 °C
Density standard, 1.6218 mg/L, 20 °C, 60 mL
Supplier: VWR Chemicals
VWR®, Density Standards, Certified Reference Materials, Referenced at 20 °C
Catalog # 99652.600
Supplier: VWR Chemicals
CAS Number:
Jump to:
Product Details & Documents
VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature.
- ISO 17025 / ISO 17034 certified under UKAS accreditation
- Density tested in strict accordance with ASTM D1480, a primary method for density determination
- Density standards range between 0,6654 - 1,6300 g/ml
- Ensures full compliance to ASTM and IP test method protocols
- Shelf life between 12 / 24 months
Specifications
Specifications
- Pk:60 mL
- Reference temperature:20 °C
- Description:Density standard
- Density:1.6218 mg/L
- Cat. No.:99652.600
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



